accessibility menu, dialog, popup

UserName

CH3OHNONN

CAS RN 4509-90-4

IUPAC Name: N-[2-(pyridin-4-yl)-1,3-benzoxazol-5-yl]acetamide
Synonyms: N-[2-(pyridin-4-yl)-1,3-benzoxazol-5-yl]acetamide
Molecular Weight (g/mol): 253.26
Molecular Formula: C14H11N3O2
InChi Key: QANQQXSQVQYQFI-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=C2OC(=NC2=C1)C1=CC=NC=C1
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)