missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromovaleryl Chloride 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 118°C |
| Quantity | 25 g |
| UN Number | 3265 |
| Formula Weight | 199.47 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Liquid |
| Chemical Name or Material | 5-Bromovaleryl Chloride |
Safety and Handling
| TSCA | No |