missing translation for 'onlineSavingsMsg'
Learn More

5-Bromoresorcinol 98.0+%, TCI America™

Catalog No. B37311G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37311G 1 g
B37315G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B37311G Supplier TCI America Supplier No. B37311G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 106120-04-1
Molecular Formula C6H5BrO2
Molecular Weight (g/mol) 189.008
MDL Number MFCD16293393
InChI Key HYHHGFFTWSYNMV-UHFFFAOYSA-N
Synonym 1-Bromo-3,5-dihydroxybenzene
PubChem CID 557383
IUPAC Name 5-bromobenzene-1,3-diol
SMILES C1=C(C=C(C=C1O)Br)O

Specifications

Melting Point 93°C
Color White-Yellow
Boiling Point 140°C
Quantity 1 g
Formula Weight 189.01
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromoresorcinol