accessibility menu, dialog, popup

UserName

OHBrOH

CAS RN 106120-04-1

IUPAC Name: 5-bromobenzene-1,3-diol
Synonyms: 5-bromobenzene-1,3-diol
Molecular Weight (g/mol): 189.01
Molecular Formula: C6H5BrO2
InChi Key: HYHHGFFTWSYNMV-UHFFFAOYSA-N
SMILES: OC1=CC(Br)=CC(O)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results