missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(5-Bromopentyl)benzene 98.0+%, TCI America™
Supplier: TCI America P132325G
Specifications
| (5-Bromopentyl)benzene | |
| Yellow | |
| C11H15Br | |
| 25 g | |
| QICUPOFVENZWSC-UHFFFAOYSA-N | |
| 5-bromopentylbenzene | |
| 285561 | |
| ≥98.0% (GC) |
| 14469-83-1 | |
| 148°C | |
| MFCD01075177 | |
| 5-bromopentyl benzene, 1-bromo-5-phenylpentane, benzene, 5-bromopentyl, 5-phenylpentyl bromide, 1-bromo-5-phenyl-pentane, 5-bromo-n-pentyl benzene, 5-bromo-pentyl-benzene, 5-brompentyl benzol, phenylpentyl bromide, 5-phenylpentylbromide | |
| C1=CC=C(C=C1)CCCCCBr | |
| 227.145 | |
| 227.15 | |
| Liquid |
Chemical Identifiers
| 14469-83-1 | |
| 227.145 | |
| QICUPOFVENZWSC-UHFFFAOYSA-N | |
| 285561 | |
| C1=CC=C(C=C1)CCCCCBr |
| C11H15Br | |
| MFCD01075177 | |
| 5-bromopentyl benzene, 1-bromo-5-phenylpentane, benzene, 5-bromopentyl, 5-phenylpentyl bromide, 1-bromo-5-phenyl-pentane, 5-bromo-n-pentyl benzene, 5-bromo-pentyl-benzene, 5-brompentyl benzol, phenylpentyl bromide, 5-phenylpentylbromide | |
| 5-bromopentylbenzene |
Safety and Handling
TSCA : No