missing translation for 'onlineSavingsMsg'
Learn More

(5-Bromopentyl)benzene 98.0+%, TCI America™

Catalog No. p-7135077
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P13235G 5 g
P132325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P13235G Supplier TCI America Supplier No. P13235G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14469-83-1
Molecular Formula C11H15Br
Molecular Weight (g/mol) 227.145
MDL Number MFCD01075177
InChI Key QICUPOFVENZWSC-UHFFFAOYSA-N
Synonym 5-bromopentyl benzene, 1-bromo-5-phenylpentane, benzene, 5-bromopentyl, 5-phenylpentyl bromide, 1-bromo-5-phenyl-pentane, 5-bromo-n-pentyl benzene, 5-bromo-pentyl-benzene, 5-brompentyl benzol, phenylpentyl bromide, 5-phenylpentylbromide
PubChem CID 285561
IUPAC Name 5-bromopentylbenzene
SMILES C1=CC=C(C=C1)CCCCCBr

Specifications

Color Yellow
Boiling Point 148°C
Quantity 5 g
Formula Weight 227.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (5-Bromopentyl)benzene