missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromoisatin 97.0+%, TCI America™
Supplier: TCI America B1566250G
Specifications
| 5-Bromoisatin | |
| 253°C | |
| MFCD00005719 | |
| 5-bromoisatin, 5-bromoindoline-2,3-dione, 5-bromoindole-2,3-dione, isatin, 5-bromo, 5-bromisatin, 1h-indole-2,3-dione, 5-bromo, 5-bromisatin czech, 5-bromo-2,3-dihydro-1h-indole-2,3-dione, indole-2,3-dione, 5-bromo, 5-bromoisatin monohydrate | |
| C1=CC2=C(C=C1Br)C(=O)C(=O)N2 | |
| 226.029 | |
| 226.03 | |
| Crystalline Powder |
| 87-48-9 | |
| C8H4BrNO2 | |
| 250 g | |
| MBVCESWADCIXJN-UHFFFAOYSA-N | |
| 5-bromo-1H-indole-2,3-dione | |
| 6889 | |
| ≥97.0% (HPLC,T) |
Chemical Identifiers
| 87-48-9 | |
| 226.029 | |
| MBVCESWADCIXJN-UHFFFAOYSA-N | |
| 6889 | |
| C1=CC2=C(C=C1Br)C(=O)C(=O)N2 |
| C8H4BrNO2 | |
| MFCD00005719 | |
| 5-bromoisatin, 5-bromoindoline-2,3-dione, 5-bromoindole-2,3-dione, isatin, 5-bromo, 5-bromisatin, 1h-indole-2,3-dione, 5-bromo, 5-bromisatin czech, 5-bromo-2,3-dihydro-1h-indole-2,3-dione, indole-2,3-dione, 5-bromo, 5-bromoisatin monohydrate | |
| 5-bromo-1H-indole-2,3-dione |
Safety and Handling
RTECSNumber : NL7875000
TSCA : No