missing translation for 'onlineSavingsMsg'
Learn More

5-Bromoisatin 97.0+%, TCI America™

Catalog No. p-7119122
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B156625G 25 g
B1566250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B156625G Supplier TCI America Supplier No. B156625G
Only null left

Chemical Identifiers

CAS 87-48-9
Molecular Formula C8H4BrNO2
Molecular Weight (g/mol) 226.029
MDL Number MFCD00005719
InChI Key MBVCESWADCIXJN-UHFFFAOYSA-N
Synonym 5-bromoisatin, 5-bromoindoline-2,3-dione, 5-bromoindole-2,3-dione, isatin, 5-bromo, 5-bromisatin, 1h-indole-2,3-dione, 5-bromo, 5-bromisatin czech, 5-bromo-2,3-dihydro-1h-indole-2,3-dione, indole-2,3-dione, 5-bromo, 5-bromoisatin monohydrate
PubChem CID 6889
IUPAC Name 5-bromo-1H-indole-2,3-dione
SMILES C1=CC2=C(C=C1Br)C(=O)C(=O)N2

Specifications

Melting Point 253°C
Quantity 25 g
Formula Weight 226.03
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromoisatin
RTECSNumber NL7875000
TSCA No