accessibility menu, dialog, popup

UserName

CAS RN 87-48-9

IUPAC Name: 5-bromo-2,3-dihydro-1H-indole-2,3-dione
Synonyms: 5-bromoisatin
Molecular Weight (g/mol): 226.03
Molecular Formula: C8H4BrNO2
InChi Key: MBVCESWADCIXJN-UHFFFAOYSA-N
SMILES: BrC1=CC=C2NC(=O)C(=O)C2=C1
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)