missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-o-anisaldehyde 98.0+%, TCI America™
$142.46 - $872.10
Chemical Identifiers
| CAS | 25016-01-7 |
|---|---|
| Molecular Formula | C8H7BrO2 |
| Molecular Weight (g/mol) | 215.046 |
| MDL Number | MFCD00003347 |
| InChI Key | IJIBRSFAXRFPPN-UHFFFAOYSA-N |
| Synonym | 5-bromo-2-anisaldehyde, 5-bromo-o-anisaldehyde, 2-methoxy-5-bromobenzaldehyde, benzaldehyde, 5-bromo-2-methoxy, 5-bromo-2-methoxy-benzaldehyde, pubchem2644, acmc-209ggg, intermediates-zcf02672, o-anisaldehyde, 5-bromo, ksc494q2j |
| PubChem CID | 90684 |
| IUPAC Name | 5-bromo-2-methoxybenzaldehyde |
| SMILES | COC1=C(C=C(C=C1)Br)C=O |
Chemical Identifiers
| 25016-01-7 | |
| 215.046 | |
| IJIBRSFAXRFPPN-UHFFFAOYSA-N | |
| 90684 | |
| COC1=C(C=C(C=C1)Br)C=O |
| C8H7BrO2 | |
| MFCD00003347 | |
| 5-bromo-2-anisaldehyde, 5-bromo-o-anisaldehyde, 2-methoxy-5-bromobenzaldehyde, benzaldehyde, 5-bromo-2-methoxy, 5-bromo-2-methoxy-benzaldehyde, pubchem2644, acmc-209ggg, intermediates-zcf02672, o-anisaldehyde, 5-bromo, ksc494q2j | |
| 5-bromo-2-methoxybenzaldehyde |
Specifications
| 25016-01-7 | |
| C8H7BrO2 | |
| 25 g | |
| IJIBRSFAXRFPPN-UHFFFAOYSA-N | |
| 5-bromo-2-methoxybenzaldehyde | |
| 90684 | |
| ≥98.0% (GC) | |
| 5-Bromo-o-anisaldehyde |
| 117°C | |
| MFCD00003347 | |
| 5-bromo-2-anisaldehyde, 5-bromo-o-anisaldehyde, 2-methoxy-5-bromobenzaldehyde, benzaldehyde, 5-bromo-2-methoxy, 5-bromo-2-methoxy-benzaldehyde, pubchem2644, acmc-209ggg, intermediates-zcf02672, o-anisaldehyde, 5-bromo, ksc494q2j | |
| COC1=C(C=C(C=C1)Br)C=O | |
| 215.046 | |
| 215.05 | |
| Crystalline Powder |
Safety and Handling
TSCA : No