missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5'-Bromo-m-terphenyl 98.0+%, TCI America™
Supplier: TCI America B430825G
Specifications
| 5′-Bromo-m-terphenyl | |
| 111°C | |
| C18H13Br | |
| 25 g | |
| 1-Bromo-3,5-diphenylbenzene, 5′-Bromo-1,1′:3′,1′′-terphenyl | |
| BrC1=CC(=CC(=C1)C1=CC=CC=C1)C1=CC=CC=C1 | |
| 309.21 | |
| 309.21 | |
| Crystalline Powder |
| 103068-20-8 | |
| White | |
| MFCD00196170 | |
| 3152 | |
| IOPQERQQZZREDR-UHFFFAOYSA-N | |
| 3-bromo-5-phenyl-1,1'-biphenyl | |
| 10070257 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 103068-20-8 | |
| 309.21 | |
| IOPQERQQZZREDR-UHFFFAOYSA-N | |
| 3-bromo-5-phenyl-1,1'-biphenyl |
| C18H13Br | |
| MFCD00196170 | |
| 10070257 | |
| BrC1=CC(=CC(=C1)C1=CC=CC=C1)C1=CC=CC=C1 |
Safety and Handling
TSCA : No