accessibility menu, dialog, popup

UserName

Br

CAS RN 103068-20-8

IUPAC Name: 3-bromo-5-phenyl-1,1'-biphenyl
Synonyms: 3-bromo-5-phenyl-1,1'-biphenyl
Molecular Weight (g/mol): 309.21
Molecular Formula: C18H13Br
InChi Key: IOPQERQQZZREDR-UHFFFAOYSA-N
SMILES: BrC1=CC(=CC(=C1)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results