missing translation for 'onlineSavingsMsg'
Learn More

5'-Bromo-m-terphenyl 98.0+%, TCI America™

Catalog No. B43085G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B43085G 5 g
B430825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B43085G Supplier TCI America Supplier No. B43085G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 103068-20-8
Molecular Formula C18H13Br
Molecular Weight (g/mol) 309.21
MDL Number MFCD00196170
InChI Key IOPQERQQZZREDR-UHFFFAOYSA-N
PubChem CID 10070257
IUPAC Name 3-bromo-5-phenyl-1,1'-biphenyl
SMILES BrC1=CC(=CC(=C1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 111°C
Color White
Quantity 5 g
UN Number 3152
Synonym 1-Bromo-3,5-diphenylbenzene, 5′C-Bromo-1,1′C:3′C,1′C′C-terphenyl
Formula Weight 309.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5′-Bromo-m-terphenyl