missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-3-nitrosalicylaldehyde 97.0+%, TCI America™

Catalog No. p-7118737
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B09081G 1 g
B09085G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B09081G Supplier TCI America Supplier No. B09081G
Only null left

Chemical Identifiers

CAS 16634-88-1
Molecular Formula C7H4BrNO4
Molecular Weight (g/mol) 246.016
MDL Number MFCD00130110
InChI Key YEYPSUQQZNDKDE-UHFFFAOYSA-N
Synonym 5-bromo-2-hydroxy-3-nitrobenzaldehyde, 5-bromo-3-nitrosalicylaldehyde, benzaldehyde, 5-bromo-2-hydroxy-3-nitro, 3-nitro-5-bromosalicylaldehyde, benzaldehyde,5-bromo-2-hydroxy-3-nitro, salicylaldehyde, 5-bromo-3-nitro, acmc-209du3, 5-bromo-3-nitrosalicyladehyde, 2-hydroxy-3-nitro-5-bromobenzaldehyde
PubChem CID 618699
SMILES C1=C(C=C(C(=C1[N+](=O)[O-])O)C=O)Br

Specifications

Melting Point 125°C
Color Yellow
Quantity 1 g
Formula Weight 246.02
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-3-nitrosalicylaldehyde