accessibility menu, dialog, popup

UserName

CAS RN 16634-88-1

IUPAC Name: 5-bromo-2-hydroxy-3-nitrobenzaldehyde
Synonyms: 5-bromo-2-hydroxy-3-nitrobenzaldehyde
Molecular Weight (g/mol): 246.02
Molecular Formula: C7H4BrNO4
InChi Key: YEYPSUQQZNDKDE-UHFFFAOYSA-N
SMILES: OC1=C(C=C(Br)C=C1C=O)[N+]([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)