missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-isopropoxybenzonitrile, 97%, Thermo Scientific Chemicals

$305.40 - $305.40

Chemical Identifiers

CAS 515832-52-7
Molecular Formula C10H10BrNO
Molecular Weight (g/mol) 240.1
MDL Number MFCD02257441
InChI Key YRPPYKWHWKFYGJ-UHFFFAOYSA-N
Synonym 5-bromo-2-isopropoxybenzonitrile, 5-bromo-2-isopropoxy-benzonitrile, benzonitrile, 5-bromo-2-1-methylethoxy, 5-bromo-2-propan-2-yloxy benzonitrile, 5-bromo-2-methylethoxy benzenecarbonitrile, acmc-1awgy, 5-bromo-2 1-methylethyl oxy benzonitrile, 5-bromo-2-1-methylethyl oxy benzonitrile
PubChem CID 3262494
IUPAC Name 5-bromo-2-propan-2-yloxybenzonitrile
SMILES CC(C)OC1=C(C=C(C=C1)Br)C#N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5062703
View Documents
Thermo Scientific Chemicals
H5062703
1 g
Each for $305.40
Only null left
 
Description

Description

5-Bromo-2-isopropoxybenzonitrile is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-2-isopropoxybenzonitrile is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
Specifications

Chemical Identifiers

515832-52-7
240.1
YRPPYKWHWKFYGJ-UHFFFAOYSA-N
3262494
CC(C)OC1=C(C=C(C=C1)Br)C#N
C10H10BrNO
MFCD02257441
5-bromo-2-isopropoxybenzonitrile, 5-bromo-2-isopropoxy-benzonitrile, benzonitrile, 5-bromo-2-1-methylethoxy, 5-bromo-2-propan-2-yloxy benzonitrile, 5-bromo-2-methylethoxy benzenecarbonitrile, acmc-1awgy, 5-bromo-2 1-methylethyl oxy benzonitrile, 5-bromo-2-1-methylethyl oxy benzonitrile
5-bromo-2-propan-2-yloxybenzonitrile

Specifications

515832-52-7
MFCD02257441
UN3439
YRPPYKWHWKFYGJ-UHFFFAOYSA-N
5-bromo-2-propan-2-yloxybenzonitrile
3262494
97%
C10H10BrNO
1 g
5-bromo-2-isopropoxybenzonitrile, 5-bromo-2-isopropoxy-benzonitrile, benzonitrile, 5-bromo-2-1-methylethoxy, 5-bromo-2-propan-2-yloxy benzonitrile, 5-bromo-2-methylethoxy benzenecarbonitrile, acmc-1awgy, 5-bromo-2 1-methylethyl oxy benzonitrile, 5-bromo-2-1-methylethyl oxy benzonitrile
CC(C)OC1=C(C=C(C=C1)Br)C#N
240.1
240.1
5-Bromo-2-isopropoxybenzonitrile
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only