CAS RN 515832-52-7
IUPAC Name:
5-bromo-2-(propan-2-yloxy)benzonitrile
Synonyms:
5-bromo-2-isopropoxybenzonitrile
Molecular Weight (g/mol):
240.1
Molecular Formula:
C10H10BrNO
InChi Key:
YRPPYKWHWKFYGJ-UHFFFAOYSA-N
SMILES:
CC(C)OC1=C(C=C(Br)C=C1)C#N
1
–
1
of
1
results
1
Promotions Available
5-Bromo-2-isopropoxybenzonitrile, 97%, Thermo Scientific Chemicals
5-Bromo-2-isopropoxybenzonitrile, 97%, Thermo Scientific Chemicals
CAS: 515832-52-7 Molecular Formula: C10H10BrNO Molecular Weight (g/mol): 240.1 MDL Number: MFCD02257441 InChI Key: YRPPYKWHWKFYGJ-UHFFFAOYSA-N Synonym: 5-bromo-2-isopropoxybenzonitrile,5-bromo-2-isopropoxy-benzonitrile,benzonitrile, 5-bromo-2-1-methylethoxy,5-bromo-2-propan-2-yloxy benzonitrile,5-bromo-2-methylethoxy benzenecarbonitrile,acmc-1awgy,5-bromo-2 1-methylethyl oxy benzonitrile,5-bromo-2-1-methylethyl oxy benzonitrile PubChem CID: 3262494 IUPAC Name: 5-bromo-2-propan-2-yloxybenzonitrile SMILES: CC(C)OC1=C(C=C(C=C1)Br)C#N