missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-isopropoxybenzonitrile, 97%, Thermo Scientific Chemicals

Catalog No. AAH5062703
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5062703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5062703 Supplier Thermo Scientific Chemicals Supplier No. H5062703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 515832-52-7 | Molecular Formula: C10H10BrNO | Molecular Weight: 240.1 g/mol

5-Bromo-2-isopropoxybenzonitrile is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-2-isopropoxybenzonitrile is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.

Chemical Identifiers

CAS 515832-52-7
Molecular Formula C10H10BrNO
Molecular Weight (g/mol) 240.1
MDL Number MFCD02257441
InChI Key YRPPYKWHWKFYGJ-UHFFFAOYSA-N
Synonym 5-bromo-2-isopropoxybenzonitrile, 5-bromo-2-isopropoxy-benzonitrile, benzonitrile, 5-bromo-2-1-methylethoxy, 5-bromo-2-propan-2-yloxy benzonitrile, 5-bromo-2-methylethoxy benzenecarbonitrile, acmc-1awgy, 5-bromo-2 1-methylethyl oxy benzonitrile, 5-bromo-2-1-methylethyl oxy benzonitrile
PubChem CID 3262494
IUPAC Name 5-bromo-2-propan-2-yloxybenzonitrile
SMILES CC(C)OC1=C(C=C(C=C1)Br)C#N

Specifications

Quantity 1 g
UN Number UN3439
Formula Weight 240.1
Percent Purity 97%
Chemical Name or Material 5-Bromo-2-isopropoxybenzonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only