missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-iodotoluene 98.0+%, TCI America™

Catalog No. p-7119460
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B20695G 5 g
B206925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B20695G Supplier TCI America Supplier No. B20695G
Only null left

Chemical Identifiers

CAS 116632-39-4
Molecular Formula C7H6BrI
Molecular Weight (g/mol) 296.93
MDL Number MFCD00060664
InChI Key GHTUADBHTFHMNI-UHFFFAOYSA-N
Synonym 5-bromo-2-iodotoluene, 1-bromo-4-iodo-5-methylbenzene, benzene, 4-bromo-1-iodo-2-methyl, bromoiodotoluene, pubchem3793, 2-iodo-5-bromo-toluene, 5-bromo-2-iodo toluene, 5-bromo-2-iodotoluene,, 4-bromo-2-methyliodobenzene, 4-bromo-2-methyl-iodobenzene
PubChem CID 2724601
IUPAC Name 4-bromo-1-iodo-2-methylbenzene
SMILES CC1=CC(Br)=CC=C1I

Specifications

Color Yellow
Boiling Point 260°C
Quantity 5 g
Formula Weight 296.93
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Bromo-2-iodotoluene