accessibility menu, dialog, popup

UserName

CAS RN 116632-39-4

IUPAC Name: 4-bromo-1-iodo-2-methylbenzene
Synonyms: 4-bromo-1-iodo-2-methylbenzene
Molecular Weight (g/mol): 296.93
Molecular Formula: C7H6BrI
InChi Key: GHTUADBHTFHMNI-UHFFFAOYSA-N
SMILES: CC1=CC(Br)=CC=C1I
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)