missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-iodotoluene, 98+%

Catalog No. AAB2187814
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2187814 25 g
AAB2187822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2187814 Supplier Thermo Scientific Chemicals Supplier No. B2187814

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 116632-39-4 | C7H6BrI | 296.93 g/mol

5-Bromo-2-iodotoluene is used as a medical intermediate and in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-2-iodotoluene is used as a medical intermediate and in organic synthesis.

Notes
Light sensitive. Incompatible with Strong oxidizing agents, light.

Chemical Identifiers

CAS 116632-39-4
Molecular Formula C7H6BrI
Molecular Weight (g/mol) 296.93
MDL Number MFCD00060664
InChI Key GHTUADBHTFHMNI-UHFFFAOYSA-N
Synonym 5-bromo-2-iodotoluene, 1-bromo-4-iodo-5-methylbenzene, benzene, 4-bromo-1-iodo-2-methyl, bromoiodotoluene, pubchem3793, 2-iodo-5-bromo-toluene, 5-bromo-2-iodo toluene, 5-bromo-2-iodotoluene,, 4-bromo-2-methyliodobenzene, 4-bromo-2-methyl-iodobenzene
PubChem CID 2724601
IUPAC Name 4-bromo-1-iodo-2-methylbenzene
SMILES CC1=CC(Br)=CC=C1I

Specifications

Density 2.139
Boiling Point 262°C to 265°C
Flash Point >110°C (230°F)
Refractive Index 1.6515
Quantity 25 g
Beilstein 2325157
Sensitivity Light sensitive
Formula Weight 296.93
Percent Purity ≥98%
Chemical Name or Material 5-Bromo-2-iodotoluene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 629-570-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only