missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-iodobenzonitrile 98.0+%, TCI America™

Catalog No. B22155G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B22155G 5 g
B22151G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B22155G Supplier TCI America Supplier No. B22155G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 121554-10-7
Molecular Formula C7H3BrIN
Molecular Weight (g/mol) 307.916
MDL Number MFCD00672950
InChI Key JIGKPMMZNMQXDL-UHFFFAOYSA-N
Synonym benzonitrile, 5-bromo-2-iodo, 5-bromo-2-iodobenzenecarbonitrile, 2-iodo-5-bromobenzonitrile, 2-cyano-5-bromo-1-iodobenzene, 5-bromo-2-iodobenzonitrile, 98+%, pubchem3790, acmc-1cir6, 5-bromo-2-iodo-benzonitrile, ksc493s3b, benzonitrile,5-bromo-2-iodo
PubChem CID 7020874
IUPAC Name 5-bromo-2-iodobenzonitrile
SMILES C1=CC(=C(C=C1Br)C#N)I

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 307.92
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-iodobenzonitrile