accessibility menu, dialog, popup

UserName

CAS RN 121554-10-7

IUPAC Name: 5-bromo-2-iodobenzonitrile
Synonyms: 5-bromo-2-iodobenzonitrile
Molecular Weight (g/mol): 307.92
Molecular Formula: C7H3BrIN
InChi Key: JIGKPMMZNMQXDL-UHFFFAOYSA-N
SMILES: BrC1=CC(C#N)=C(I)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)