missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-fluoro-m-xylene 97.0+%, TCI America™

Catalog No. B43701G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B43701G 1 g
B43705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B43701G Supplier TCI America Supplier No. B43701G

Chemical Identifiers

CAS 99725-44-7
Molecular Formula C8H8BrF
Molecular Weight (g/mol) 203.054
MDL Number MFCD01320701
InChI Key ZXPHUVHMBKRRJF-UHFFFAOYSA-N
Synonym 5-bromo-2-fluoro-m-xylene, 5-bromo-2-fluoro-3-xylene, 4-bromo-2,6-dimethylfluorobenzene, 1-bromo-4-fluoro-3,5-dimethylbenzene, 5-bromo-2-fluoro-1,3-xylene, benzene, 5-bromo-2-fluoro-1,3-dimethyl, 3,5-dimethyl-4-fluorobromobenzene, 5-bromo-2-fluoro-1,3-dimethyl-benzene, ksc486i9j
PubChem CID 2736298
IUPAC Name 5-bromo-2-fluoro-1,3-dimethylbenzene
SMILES CC1=CC(=CC(=C1F)C)Br

Specifications

Color Yellow
Quantity 1 g
Physical Form Clear Liquid at 20°C
Boiling Point 89°C
Formula Weight 203.05
Percent Purity ≥97.0% (GC)
Chemical Name or Material 5-Bromo-2-fluoro-m-xylene