accessibility menu, dialog, popup

UserName

CAS RN 99725-44-7

CH3FH3CBr

CAS RN 99725-44-7

IUPAC Name: 5-bromo-2-fluoro-1,3-dimethylbenzene
Synonyms: 5-bromo-2-fluoro-1,3-dimethylbenzene
Molecular Weight (g/mol): 203.05
Molecular Formula: C8H8BrF
InChi Key: ZXPHUVHMBKRRJF-UHFFFAOYSA-N
SMILES: CC1=CC(Br)=CC(C)=C1F
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)