missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-chlorobenzyl Alcohol 98.0+%, TCI America™

$111.66 - $350.38

Chemical Identifiers

CAS 149965-40-2
Molecular Formula C7H6BrClO
Molecular Weight (g/mol) 221.48
MDL Number MFCD02683548
InChI Key SCHBQPIVMBDOQF-UHFFFAOYSA-N
PubChem CID 5074766
IUPAC Name (5-bromo-2-chlorophenyl)methanol
SMILES OCC1=C(Cl)C=CC(Br)=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B39461G
View Documents
TCI America
B39461G
1 g
Each for $111.66
Only null left
 
B39465G
View Documents
TCI America
B39465G
5 g
Each for $350.38
Only null left
 
Description

Description

Specifications

Chemical Identifiers

149965-40-2
221.48
SCHBQPIVMBDOQF-UHFFFAOYSA-N
(5-bromo-2-chlorophenyl)methanol
C7H6BrClO
MFCD02683548
5074766
OCC1=C(Cl)C=CC(Br)=C1

Specifications

149965-40-2
White-Yellow
MFCD02683548
SCHBQPIVMBDOQF-UHFFFAOYSA-N
(5-bromo-2-chlorophenyl)methanol
5074766
≥98.0% (GC)
5-Bromo-2-chlorobenzyl Alcohol
94°C
C7H6BrClO
1 g
OCC1=C(Cl)C=CC(Br)=C1
221.48
221.48
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'