accessibility menu, dialog, popup

UserName

BrOHCl

CAS RN 149965-40-2

IUPAC Name: (5-bromo-2-chlorophenyl)methanol
Synonyms: (5-bromo-2-chlorophenyl)methanol
Molecular Weight (g/mol): 221.48
Molecular Formula: C7H6BrClO
InChi Key: SCHBQPIVMBDOQF-UHFFFAOYSA-N
SMILES: OCC1=C(Cl)C=CC(Br)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results