missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-chlorobenzyl Alcohol 98.0+%, TCI America™

Catalog No. B39465G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B39465G 5 g
B39461G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B39465G Supplier TCI America Supplier No. B39465G
Only null left

Chemical Identifiers

CAS 149965-40-2
Molecular Formula C7H6BrClO
Molecular Weight (g/mol) 221.48
MDL Number MFCD02683548
InChI Key SCHBQPIVMBDOQF-UHFFFAOYSA-N
PubChem CID 5074766
IUPAC Name (5-bromo-2-chlorophenyl)methanol
SMILES OCC1=C(Cl)C=CC(Br)=C1

Specifications

Melting Point 94°C
Color White-Yellow
Quantity 5 g
Formula Weight 221.48
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-chlorobenzyl Alcohol