missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-chlorobenzoic Acid 98.0+%, TCI America™

Catalog No. B134425G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B134425G 25 g
B134410G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B134425G Supplier TCI America Supplier No. B134425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 21739-92-4
Molecular Formula C7H4BrClO2
Molecular Weight (g/mol) 235.461
MDL Number MFCD00002415
InChI Key FGERXQWKKIVFQG-UHFFFAOYSA-N
Synonym 2-chloro-5-bromobenzoic acid, benzoic acid, 5-bromo-2-chloro, 5-bromo-2-chlorobenzoicacid, 5-bromo-2-chloro benzoic acid, 5-bromo-2-chloro-benzoic acid, bromo 5--2-chlorobenzoic acid, pubchem3587, acmc-209fnm, 5-bromo-2-chorobenzoic acid, ksc204k3r
PubChem CID 33127
IUPAC Name 5-bromo-2-chlorobenzoic acid
SMILES C1=CC(=C(C=C1Br)C(=O)O)Cl

Specifications

Melting Point 158°C
Color White-Yellow
Quantity 25 g
Formula Weight 235.46
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-chlorobenzoic Acid