accessibility menu, dialog, popup

UserName

OOBrCl

CAS RN 21739-92-4

IUPAC Name: 5-bromo-2-chlorobenzoate
Synonyms: 5-bromo-2-chlorobenzoate
Molecular Weight (g/mol): 234.45
Molecular Formula: C7H3BrClO2
InChi Key: FGERXQWKKIVFQG-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC(Br)=CC=C1Cl
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)