missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-chlorobenzoic acid, 98+%

Catalog No. AAA1221809
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1221809 10 g
AAA1221818 50 g
AAA1221830 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1221809 Supplier Thermo Scientific Chemicals Supplier No. A1221809
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 21739-92-4 | C7H4BrClO2 | 235.461 g/mol

5-Bromo-2-chlorobenzoic acid was used to evaluate the homogeneity of organic powder. It can be used to produce 2-chloro-5-deuteriobenzoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-2-chlorobenzoic acid was used to evaluate the homogeneity of organic powder. It can be used to produce 2-chloro-5-deuteriobenzoic acid.

Solubility
Soluble in water 2.63 g/L @20°C.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 21739-92-4
Molecular Formula C7H4BrClO2
Molecular Weight (g/mol) 235.461
MDL Number MFCD00002415
InChI Key FGERXQWKKIVFQG-UHFFFAOYSA-N
Synonym 2-chloro-5-bromobenzoic acid, benzoic acid, 5-bromo-2-chloro, 5-bromo-2-chlorobenzoicacid, 5-bromo-2-chloro benzoic acid, 5-bromo-2-chloro-benzoic acid, bromo 5--2-chlorobenzoic acid, pubchem3587, acmc-209fnm, 5-bromo-2-chorobenzoic acid, ksc204k3r
PubChem CID 33127
IUPAC Name 5-bromo-2-chlorobenzoic acid
SMILES C1=CC(=C(C=C1Br)C(=O)O)Cl

Specifications

Melting Point 157°C to 162°C
Odor Odorless
Quantity 10 g
Beilstein 2691432
Solubility Information Soluble in water 2.63g/L @20°C.
Formula Weight 235.47
Percent Purity ≥98%
Chemical Name or Material 5-Bromo-2-chlorobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 244-558-5
TSCA Yes

RUO – Research Use Only