missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-2-chlorobenzaldehyde 98.0+%, TCI America™
$46.20 - $130.92
Chemical Identifiers
| CAS | 189628-37-3 |
|---|---|
| Molecular Formula | C7H4BrClO |
| Molecular Weight (g/mol) | 219.46 |
| MDL Number | MFCD08445659 |
| InChI Key | DPKKRQAEYWOISP-UHFFFAOYSA-N |
| PubChem CID | 10608925 |
| IUPAC Name | 5-bromo-2-chlorobenzaldehyde |
| SMILES | ClC1=C(C=O)C=C(Br)C=C1 |
Chemical Identifiers
| 189628-37-3 | |
| 219.46 | |
| DPKKRQAEYWOISP-UHFFFAOYSA-N | |
| 5-bromo-2-chlorobenzaldehyde |
| C7H4BrClO | |
| MFCD08445659 | |
| 10608925 | |
| ClC1=C(C=O)C=C(Br)C=C1 |
Specifications
| 189628-37-3 | |
| Yellow | |
| MFCD08445659 | |
| 2-chloro-5-bromobenzaldehyde, 5-bromo-2-chloro-benzaldehyde, benzaldehyde, 5-bromo-2-chloro, pubchem10373, acmc-1bzt6, ksc498g6h, 5-bromo-2-chloro benzaldehyde, benzaldehyde,5-bromo-2-chloro, 5-bromanyl-2-chloranyl-benzaldehyde | |
| ClC1=C(C=O)C=C(Br)C=C1 | |
| 219.46 | |
| 219.46 | |
| Crystalline Powder |
| 71°C | |
| C7H4BrClO | |
| 1 g | |
| DPKKRQAEYWOISP-UHFFFAOYSA-N | |
| 5-bromo-2-chlorobenzaldehyde | |
| 10608925 | |
| ≥98.0% (GC) | |
| 5-Bromo-2-chlorobenzaldehyde |
Safety and Handling
TSCA : No