accessibility menu, dialog, popup

UserName

BrOCl

CAS RN 189628-37-3

IUPAC Name: 5-bromo-2-chlorobenzaldehyde
Synonyms: 5-bromo-2-chlorobenzaldehyde
Molecular Weight (g/mol): 219.46
Molecular Formula: C7H4BrClO
InChi Key: DPKKRQAEYWOISP-UHFFFAOYSA-N
SMILES: ClC1=C(C=O)C=C(Br)C=C1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)