missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-chlorobenzaldehyde 98.0+%, TCI America™

Catalog No. B47565G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B47565G 5 g
B47561G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B47565G Supplier TCI America Supplier No. B47565G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 189628-37-3
Molecular Formula C7H4BrClO
Molecular Weight (g/mol) 219.46
MDL Number MFCD08445659
InChI Key DPKKRQAEYWOISP-UHFFFAOYSA-N
PubChem CID 10608925
IUPAC Name 5-bromo-2-chlorobenzaldehyde
SMILES ClC1=C(C=O)C=C(Br)C=C1

Specifications

Melting Point 71°C
Color Yellow
Quantity 5 g
Synonym 2-chloro-5-bromobenzaldehyde, 5-bromo-2-chloro-benzaldehyde, benzaldehyde, 5-bromo-2-chloro, pubchem10373, acmc-1bzt6, ksc498g6h, 5-bromo-2-chloro benzaldehyde, benzaldehyde,5-bromo-2-chloro, 5-bromanyl-2-chloranyl-benzaldehyde
Formula Weight 219.46
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-chlorobenzaldehyde