missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-2-chlorobenzaldehyde 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 71°C |
| Color | Yellow |
| Quantity | 1 g |
| Synonym | 2-chloro-5-bromobenzaldehyde, 5-bromo-2-chloro-benzaldehyde, benzaldehyde, 5-bromo-2-chloro, pubchem10373, acmc-1bzt6, ksc498g6h, 5-bromo-2-chloro benzaldehyde, benzaldehyde,5-bromo-2-chloro, 5-bromanyl-2-chloranyl-benzaldehyde |
| Formula Weight | 219.46 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 5-Bromo-2-chlorobenzaldehyde |
Safety and Handling
| TSCA | No |