missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-chlorobenzaldehyde, 98%, Thermo Scientific Chemicals

$82.44 - $363.66

Chemical Identifiers

CAS 189628-37-3
Molecular Formula C7H4BrClO
Molecular Weight (g/mol) 219.46
MDL Number MFCD08445659
InChI Key DPKKRQAEYWOISP-UHFFFAOYSA-N
PubChem CID 10608925
IUPAC Name 5-bromo-2-chlorobenzaldehyde
SMILES ClC1=C(C=O)C=C(Br)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6443803
View Documents
Thermo Scientific Chemicals
H6443803
1 g
Each for $82.44
Only null left
 
AAH6443806
View Documents
Thermo Scientific Chemicals
H6443806
5 g
Each for $363.66
Only null left
 
Description

Description

It finds its application as an intermediate in organic syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application as an intermediate in organic syntheses.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

189628-37-3
219.46
DPKKRQAEYWOISP-UHFFFAOYSA-N
5-bromo-2-chlorobenzaldehyde
C7H4BrClO
MFCD08445659
10608925
ClC1=C(C=O)C=C(Br)C=C1

Specifications

189628-37-3
C7H4BrClO
1 g
2-chloro-5-bromobenzaldehyde, 5-bromo-2-chloro-benzaldehyde, benzaldehyde, 5-bromo-2-chloro, pubchem10373, acmc-1bzt6, ksc498g6h, 5-bromo-2-chloro benzaldehyde, benzaldehyde,5-bromo-2-chloro, 5-bromanyl-2-chloranyl-benzaldehyde
DPKKRQAEYWOISP-UHFFFAOYSA-N
5-bromo-2-chlorobenzaldehyde
10608925
98%
5-Bromo-2-chlorobenzaldehyde
White to Yellow
MFCD08445659
9757388
Slightly soluble in water.
ClC1=C(C=O)C=C(Br)C=C1
219.46
219.46
Solid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only