missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-chlorobenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog No. AAH6443803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6443803 1 g
AAH6443806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6443803 Supplier Thermo Scientific Chemicals Supplier No. H6443803

It finds its application as an intermediate in organic syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application as an intermediate in organic syntheses.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 189628-37-3
Molecular Formula C7H4BrClO
Molecular Weight (g/mol) 219.46
MDL Number MFCD08445659
InChI Key DPKKRQAEYWOISP-UHFFFAOYSA-N
PubChem CID 10608925
IUPAC Name 5-bromo-2-chlorobenzaldehyde
SMILES ClC1=C(C=O)C=C(Br)C=C1

Specifications

Color White to Yellow
Quantity 1 g
Beilstein 9757388
Synonym 2-chloro-5-bromobenzaldehyde, 5-bromo-2-chloro-benzaldehyde, benzaldehyde, 5-bromo-2-chloro, pubchem10373, acmc-1bzt6, ksc498g6h, 5-bromo-2-chloro benzaldehyde, benzaldehyde,5-bromo-2-chloro, 5-bromanyl-2-chloranyl-benzaldehyde
Solubility Information Slightly soluble in water.
Formula Weight 219.46
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 5-Bromo-2-chlorobenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only