missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-chloroanisole 98.0+%, TCI America™

Catalog No. B30011G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B30011G 1 g
B30015G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B30011G Supplier TCI America Supplier No. B30011G

Chemical Identifiers

CAS 16817-43-9
Molecular Formula C7H6BrClO
Molecular Weight (g/mol) 221.48
MDL Number MFCD00078659
InChI Key UAMVKOTWSHJOSY-UHFFFAOYSA-N
Synonym 5-bromo-2-chloroanisole, 5-bromo-2-chloroanisol, 4-chloro-3-methoxybromobenzene, 2-chloro-5-bromoanisole, 5-bromo-2-chloro-anisole, benzene, 4-bromo-1-chloro-2-methoxy, 4-bromo-1-chloro-2-methoxy-benzene, zlchem 582, pubchem2632, acmc-209dxq
PubChem CID 2769567
IUPAC Name 4-bromo-1-chloro-2-methoxybenzene
SMILES COC1=C(Cl)C=CC(Br)=C1

Specifications

Color White-Yellow
Boiling Point 130°C
Quantity 1 g
Formula Weight 221.48
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material 5-Bromo-2-chloroanisole