accessibility menu, dialog, popup

UserName

H3COBrCl

CAS RN 16817-43-9

IUPAC Name: 4-bromo-1-chloro-2-methoxybenzene
Synonyms: 4-bromo-1-chloro-2-methoxybenzene
Molecular Weight (g/mol): 221.48
Molecular Formula: C7H6BrClO
InChi Key: UAMVKOTWSHJOSY-UHFFFAOYSA-N
SMILES: COC1=C(Cl)C=CC(Br)=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)