missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-anisaldehyde, 99%, Thermo Scientific™

Chemical Identifiers

CAS 25016-01-7
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.05
MDL Number MFCD00003347
InChI Key IJIBRSFAXRFPPN-UHFFFAOYSA-N
Synonym 5-bromo-2-anisaldehyde, 5-bromo-o-anisaldehyde, 2-methoxy-5-bromobenzaldehyde, benzaldehyde, 5-bromo-2-methoxy, 5-bromo-2-methoxy-benzaldehyde, pubchem2644, acmc-209ggg, intermediates-zcf02672, o-anisaldehyde, 5-bromo, ksc494q2j
PubChem CID 90684
IUPAC Name 5-bromo-2-methoxybenzaldehyde
SMILES COC1=C(C=C(C=C1)Br)C=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC167520050
SDS View Documents
Thermo Scientific Chemicals
167520050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC167520250
SDS View Documents
Thermo Scientific Chemicals
167520250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC167521000
SDS View Documents
Thermo Scientific Chemicals
167521000
100g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

25016-01-7
215.05
IJIBRSFAXRFPPN-UHFFFAOYSA-N
90684
COC1=C(C=C(C=C1)Br)C=O
C8H7BrO2
MFCD00003347
5-bromo-2-anisaldehyde, 5-bromo-o-anisaldehyde, 2-methoxy-5-bromobenzaldehyde, benzaldehyde, 5-bromo-2-methoxy, 5-bromo-2-methoxy-benzaldehyde, pubchem2644, acmc-209ggg, intermediates-zcf02672, o-anisaldehyde, 5-bromo, ksc494q2j
5-bromo-2-methoxybenzaldehyde

Specifications

25016-01-7
100.0
98.5% min. (GC)
C8H7BrO2
MFCD00003347
08, 55
IJIBRSFAXRFPPN-UHFFFAOYSA-N
5-bromo-2-methoxybenzaldehyde
90684
99%
98.5
Authentic
Glass bottle
BrC6H3(OCH3)CHO
5g
5-bromo-2-anisaldehyde, 5-bromo-o-anisaldehyde, 2-methoxy-5-bromobenzaldehyde, benzaldehyde, 5-bromo-2-methoxy, 5-bromo-2-methoxy-benzaldehyde, pubchem2644, acmc-209ggg, intermediates-zcf02672, o-anisaldehyde, 5-bromo, ksc494q2j
COC1=C(C=C(C=C1)Br)C=O
215.05
215.05
5-Bromo-2-anisaldehyde
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attent

GHS Signal Word: Warning

EINECSNumber : 246-564-3

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only