missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-anisaldehyde, 99%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 167520250

 View more versions of this product

Catalog No. AC167520250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

5-Bromo-2-anisaldehyde
98.5
Authentic
Glass bottle
BrC6H3(OCH3)CHO
25g
5-bromo-2-anisaldehyde, 5-bromo-o-anisaldehyde, 2-methoxy-5-bromobenzaldehyde, benzaldehyde, 5-bromo-2-methoxy, 5-bromo-2-methoxy-benzaldehyde, pubchem2644, acmc-209ggg, intermediates-zcf02672, o-anisaldehyde, 5-bromo, ksc494q2j
COC1=C(C=C(C=C1)Br)C=O
215.05
215.05
25016-01-7
100.0
98.5% min. (GC)
C8H7BrO2
MFCD00003347
08, 55
IJIBRSFAXRFPPN-UHFFFAOYSA-N
5-bromo-2-methoxybenzaldehyde
90684
99%

Chemical Identifiers

25016-01-7
215.05
IJIBRSFAXRFPPN-UHFFFAOYSA-N
90684
COC1=C(C=C(C=C1)Br)C=O
C8H7BrO2
MFCD00003347
5-bromo-2-anisaldehyde, 5-bromo-o-anisaldehyde, 2-methoxy-5-bromobenzaldehyde, benzaldehyde, 5-bromo-2-methoxy, 5-bromo-2-methoxy-benzaldehyde, pubchem2644, acmc-209ggg, intermediates-zcf02672, o-anisaldehyde, 5-bromo, ksc494q2j
5-bromo-2-methoxybenzaldehyde
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attent

GHS Signal Word: Warning

EINECSNumber : 246-564-3

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only