missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2,4-difluorotoluene, 98%

Catalog No. p-7031176
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3215506 5 g
AAH3215514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3215506 Supplier Thermo Scientific Chemicals Supplier No. H3215506

CAS: 159277-47-1 | C7H5BrF2 | 207.018 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 159277-47-1
Molecular Formula C7H5BrF2
Molecular Weight (g/mol) 207.018
MDL Number MFCD15144620
InChI Key WOCZKGUFVNUBEC-UHFFFAOYSA-N
Synonym 5-bromo-2,4-difluorotoluene, benzene, 1-bromo-2,4-difluoro-5-methyl, 1-methyl-2,4-difluoro-5-bromobenzene, 2,4-difluoro-5-bromo toluene
PubChem CID 10081748
IUPAC Name 1-bromo-2,4-difluoro-5-methylbenzene
SMILES CC1=CC(=C(C=C1F)F)Br

Specifications

Quantity 5 g
Formula Weight 207.02
Percent Purity 98%
Chemical Name or Material 5-Bromo-2,4-difluorotoluene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only