CAS RN 159277-47-1
IUPAC Name:
1-bromo-2,4-difluoro-5-methylbenzene
Synonyms:
1-bromo-2,4-difluoro-5-methylbenzene
Molecular Weight (g/mol):
207.02
Molecular Formula:
C7H5BrF2
InChi Key:
WOCZKGUFVNUBEC-UHFFFAOYSA-N
SMILES:
CC1=CC(Br)=C(F)C=C1F
1
–
1
of
1
results
1
Promotions Available
5-Bromo-2,4-difluorotoluene, 98%
5-Bromo-2,4-difluorotoluene, 98%
CAS: 159277-47-1 Molecular Formula: C7H5BrF2 Molecular Weight (g/mol): 207.018 MDL Number: MFCD15144620 InChI Key: WOCZKGUFVNUBEC-UHFFFAOYSA-N Synonym: 5-bromo-2,4-difluorotoluene,benzene, 1-bromo-2,4-difluoro-5-methyl,1-methyl-2,4-difluoro-5-bromobenzene,2,4-difluoro-5-bromo toluene PubChem CID: 10081748 IUPAC Name: 1-bromo-2,4-difluoro-5-methylbenzene SMILES: CC1=CC(=C(C=C1F)F)Br