missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2,1,3-benzoxadiazole, 97%, Thermo Scientific™

Chemical Identifiers

CAS 51376-06-8
Molecular Formula C6H3BrN2O
Molecular Weight (g/mol) 199.01
MDL Number MFCD02682026
InChI Key ZWDFFESFCIACQC-UHFFFAOYSA-N
Synonym 5-bromobenzo c 1,2,5 oxadiazole, 5-bromobenzofurazan, 2,1,3-benzoxadiazole, 5-bromo, 5-bromo-benzo 1,2,5 oxadiazole, 2,1,3-benzoxadiazole,5-bromo, 5-bromobenzfurazan, pubchem15922, acmc-209ktf, amth003, buttpark 85\04-54
PubChem CID 2776298
IUPAC Name 5-bromo-2,1,3-benzoxadiazole
SMILES BrC1=CC2=NON=C2C=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
CC09710EA
View Documents
N/A 10g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

51376-06-8
199.01
ZWDFFESFCIACQC-UHFFFAOYSA-N
2776298
BrC1=CC2=NON=C2C=C1
C6H3BrN2O
MFCD02682026
5-bromobenzo c 1,2,5 oxadiazole, 5-bromobenzofurazan, 2,1,3-benzoxadiazole, 5-bromo, 5-bromo-benzo 1,2,5 oxadiazole, 2,1,3-benzoxadiazole,5-bromo, 5-bromobenzfurazan, pubchem15922, acmc-209ktf, amth003, buttpark 85\04-54
5-bromo-2,1,3-benzoxadiazole

Specifications

51376-06-8
100.0
C6H3BrN2O
5-bromobenzo c 1,2,5 oxadiazole, 5-bromobenzofurazan, 2,1,3-benzoxadiazole, 5-bromo, 5-bromo-benzo 1,2,5 oxadiazole, 2,1,3-benzoxadiazole,5-bromo, 5-bromobenzfurazan, pubchem15922, acmc-209ktf, amth003, buttpark 85\04-54
BrC1=CC2=NON=C2C=C1
199.01
199.01
5-Bromo-2, 1, 3-benzoxadiazole
96.5
97%
MFCD02682026
ZWDFFESFCIACQC-UHFFFAOYSA-N
5-bromo-2,1,3-benzoxadiazole
2776298
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents