missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2,1,3-benzoxadiazole, 97%, Thermo Scientific™

Catalog No. CC09710EA
Change view
Click to view available options
Quantity:
10g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC09710EA 10g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC09710EA Supplier Thermo Scientific Chemicals Supplier No. CC09710EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 51376-06-8
Molecular Formula C6H3BrN2O
Molecular Weight (g/mol) 199.01
MDL Number MFCD02682026
InChI Key ZWDFFESFCIACQC-UHFFFAOYSA-N
Synonym 5-bromobenzo c 1,2,5 oxadiazole, 5-bromobenzofurazan, 2,1,3-benzoxadiazole, 5-bromo, 5-bromo-benzo 1,2,5 oxadiazole, 2,1,3-benzoxadiazole,5-bromo, 5-bromobenzfurazan, pubchem15922, acmc-209ktf, amth003, buttpark 85\04-54
PubChem CID 2776298
IUPAC Name 5-bromo-2,1,3-benzoxadiazole
SMILES BrC1=CC2=NON=C2C=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10g
Formula Weight 199.01
Percent Purity 97%
Chemical Name or Material 5-Bromo-2, 1, 3-benzoxadiazole
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.