missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-1-pentene 95.0+%, TCI America™

Catalog No. p-7119071
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B14745G 5 g
B147425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B14745G Supplier TCI America Supplier No. B14745G
Only null left

Chemical Identifiers

CAS 1119-51-3
Molecular Formula C5H9Br
Molecular Weight (g/mol) 149.031
MDL Number MFCD00000264
InChI Key LPNANKDXVBMDKE-UHFFFAOYSA-N
Synonym 5-bromo-1-pentene, 1-pentene, 5-bromo, 1-bromo-4-pentene, 4-pentenyl bromide, 5-bromopentene, 5-bromo-pent-1-ene, 5-bromo-pentene, 5-bromopentene-1, 5-bromo-1-amylene, pent-4-enyl bromide
PubChem CID 70704
IUPAC Name 5-bromopent-1-ene
SMILES C=CCCCBr

Specifications

Color Yellow
Boiling Point 126°C
Quantity 5 g
UN Number 1993
Formula Weight 149.03
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Bromo-1-pentene
EINECSNumber (2)-0067
TSCA Yes
Recommended Storage Refrigerator