missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-1-methyl-1H-indole, 97%, Thermo Scientific™

Chemical Identifiers

CAS 10075-52-2
Molecular Formula C9H8BrN
Molecular Weight (g/mol) 210.07
MDL Number MFCD00159857
InChI Key SBOITLSQLQGSLO-UHFFFAOYSA-N
Synonym 5-bromo-1-methyl-1h-indole, 5-bromo-1-methyl-indole, 1h-indole, 5-bromo-1-methyl, 1-methyl-5-bromoindole, 1h-indole,5-bromo-1-methyl, pubchem7946, zlchem 1345, n-methyl-5-bromoindole, 5-bromo-n-methylindole, acmc-2097se
PubChem CID 285757
IUPAC Name 5-bromo-1-methylindole
SMILES CN1C=CC2=CC(Br)=CC=C12
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
CC41410DE
View Documents
N/A 5g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

10075-52-2
210.07
SBOITLSQLQGSLO-UHFFFAOYSA-N
285757
CN1C=CC2=CC(Br)=CC=C12
C9H8BrN
MFCD00159857
5-bromo-1-methyl-1h-indole, 5-bromo-1-methyl-indole, 1h-indole, 5-bromo-1-methyl, 1-methyl-5-bromoindole, 1h-indole,5-bromo-1-methyl, pubchem7946, zlchem 1345, n-methyl-5-bromoindole, 5-bromo-n-methylindole, acmc-2097se
5-bromo-1-methylindole

Specifications

10075-52-2
C9H8BrN
5-bromo-1-methyl-1h-indole, 5-bromo-1-methyl-indole, 1h-indole, 5-bromo-1-methyl, 1-methyl-5-bromoindole, 1h-indole,5-bromo-1-methyl, pubchem7946, zlchem 1345, n-methyl-5-bromoindole, 5-bromo-n-methylindole, acmc-2097se
CN1C=CC2=CC(Br)=CC=C12
210.07
210.07
5-Bromo-1-methyl-1H-indole
97%
MFCD00159857
SBOITLSQLQGSLO-UHFFFAOYSA-N
5-bromo-1-methylindole
285757
97%
Videos
SDS
Documents

Documents