accessibility menu, dialog, popup

UserName

H3CNBr

CAS RN 10075-52-2

IUPAC Name: 5-bromo-1-methyl-1H-indole
Synonyms: 5-bromo-1-methylindole
Molecular Weight (g/mol): 210.07
Molecular Formula: C9H8BrN
InChi Key: SBOITLSQLQGSLO-UHFFFAOYSA-N
SMILES: CN1C=CC2=CC(Br)=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)