missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-1,3-difluoro-2-(trifluoromethoxy)benzene 98.0+%, TCI America™

Catalog No. B50575G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B50575G 5 g
B50571G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B50575G Supplier TCI America Supplier No. B50575G

Chemical Identifiers

CAS 115467-07-7
Molecular Formula C7H2BrF5O
Molecular Weight (g/mol) 276.99
MDL Number MFCD03412225
InChI Key ORHCJZZKAUAZDR-UHFFFAOYSA-N
PubChem CID 15219533
IUPAC Name 5-bromo-1,3-difluoro-2-(trifluoromethoxy)benzene
SMILES FC1=CC(Br)=CC(F)=C1OC(F)(F)F

Specifications

Color Yellow
Quantity 5 g
Formula Weight 276.99
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Bromo-1,3-difluoro-2-(trifluoromethoxy)benzene