accessibility menu, dialog, popup

UserName

FOFFFFBr

CAS RN 115467-07-7

IUPAC Name: 5-bromo-1,3-difluoro-2-(trifluoromethoxy)benzene
Synonyms: 5-bromo-1,3-difluoro-2-(trifluoromethoxy)benzene
Molecular Weight (g/mol): 276.99
Molecular Formula: C7H2BrF5O
InChi Key: ORHCJZZKAUAZDR-UHFFFAOYSA-N
SMILES: FC1=CC(Br)=CC(F)=C1OC(F)(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results